Polyclonal antibody for 192519091908 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. 192519091908 information: Molecular Weight: 54537 MW; Subcellular Localization: Nucleus. Cytoplasm. Mitochondrion outer membrane. Shuttles between the cytoplasm and the nucleus and heterodimerization with JUN is essential for the nuclear localization. Localization to the cytoplasm is observed under conditions of cellular stress and in disease states. Localizes at the mitochondrial outer membrane in response to genotoxic stress. Phosphorylation at Thr-52 is required for its nuclear localization and negatively regulates its mitochondrial localization. Co- localizes with the MRN complex in the IR-induced foci (IRIF); Tissue Specificity: Ubiquitously expressed, with more abundant expression in the brain.
Type: Primary
Antigen: ATF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat