Anti-Caveolin-1 Polyclonal Antibody

Lieferant: Boster Bio
PB9165
BSBTPB9165EA 552 EUR
BSBTPB9165
Anti-Caveolin-1 Polyclonal Antibody
Antikörper
Polyclonal antibody for CAVEOLIN 1/CAV1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CAVEOLIN 1/CAV1 information: Molecular Weight: 20472 MW; Subcellular Localization: Golgi apparatus membrane; Peripheral membrane protein. Cell membrane; Peripheral membrane protein. Membrane, caveola; Peripheral membrane protein. Membrane raft. Colocalized with DPP4 in membrane rafts. Potential hairpin- like structure in the membrane. Membrane protein of caveolae; Tissue Specificity: Expressed in muscle and lung, less so in liver, brain and kidney.

Type: Primary
Antigen: Caveolin-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR