Anti-EPB41L1/4.1N Polyclonal Antibody

Lieferant: Boster Bio
PB9179
BSBTPB9179EA 552 EUR
BSBTPB9179
Anti-EPB41L1/4.1N Polyclonal Antibody
Antikörper
Polyclonal antibody for 4.1N/EPB41L1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. 4.1N/EPB41L1 information: Molecular Weight: 98503 MW; Subcellular Localization: Cytoplasm, cytoskeleton; Tissue Specificity: Highest expression in brain, lower in heart, kidney, pancreas, placenta, lung and skeletal muscle.

Type: Primary
Antigen: EPB41L1/4.1N
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR