Polyclonal antibody for EDA A2/EDA detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. EDA A2/EDA information: Molecular Weight: 41294 MW; Subcellular Localization: Cell membrane; Single-pass type II membrane protein; Tissue Specificity: Not abundant; expressed in specific cell types of ectodermal (but not mesodermal) origin of keratinocytes, hair follicles, sweat glands. Also in adult heart, liver, muscle, pancreas, prostate, fetal liver, uterus, small intestine and umbilical chord.
Type: Primary
Antigen: EDA/Eda A2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human