Anti-FOXM1 Polyclonal Antibody

Lieferant: Boster Bio
PB9194
BSBTPB9194EA 552 EUR
BSBTPB9194
Anti-FOXM1 Polyclonal Antibody
Antikörper
Polyclonal antibody for FOXM1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. FOXM1 information: Molecular Weight: 84283 MW; Subcellular Localization: Nucleus; Tissue Specificity: Expressed in thymus, testis, small intestine, colon followed by ovary. Appears to be expressed only in adult organs containing proliferating/cycling cells or in response to growth factors. Also expressed in epithelial cell lines derived from tumors. Not expressed in resting cells. Isoform 2 is highly expressed in testis.

Type: Primary
Antigen: FOXM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR