Polyclonal antibody for GLUCOCORTICOID RECEPTOR/NR3C1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. GLUCOCORTICOID RECEPTOR/NR3C1 information: Molecular Weight: 85659 MW; Subcellular Localization: Cytoplasm . Mitochondrion. Nucleus . Cytoplasmic in the absence of ligand, nuclear after ligand- binding; Tissue Specificity: Widely expressed. In the heart, detected in left and right atria, left and right ventricles, aorta, apex, intraventricular septum, and atrioventricular node as well as whole adult and fetal heart.
Type: Primary
Antigen: NR3C1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat