Anti-ZEB2/Sip1 Polyclonal Antibody

Lieferant: Boster Bio
PB9236
BSBTPB9236EA 552 EUR
BSBTPB9236
Anti-ZEB2/Sip1 Polyclonal Antibody
Antikörper
Polyclonal antibody for SIP1/ZEB2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. SIP1/ZEB2 information: Molecular Weight: 136447 MW; Subcellular Localization: Nucleus .

Type: Primary
Antigen: ZEB2/Sip1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR