Anti-Kallikrein 3 Polyclonal Antibody

Lieferant: Boster Bio
PB9259
BSBTPB9259EA 552 EUR
BSBTPB9259
Anti-Kallikrein 3 Polyclonal Antibody
Antikörper
Polyclonal antibody for PSA/KLK3 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. PSA/KLK3 information: Molecular Weight: 28741 MW; Subcellular Localization: Secreted.

Type: Primary
Antigen: Kallikrein 3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR