Polyclonal antibody for Mesothelin/MSLN detection. Host: Rabbit.Size: 100μg/vial. Tested applications: ELISA. Reactive species: Human. Mesothelin/MSLN information: Molecular Weight: 68986 MW; Subcellular Localization: Cell membrane; Lipid-anchor, GPI-anchor. Golgi apparatus; Tissue Specificity: Expressed in lung. Expressed at low levels in heart, placenta and kidney. Expressed in mesothelial cells. Highly expressed in mesotheliomas, ovarian cancers, and some squamous cell carcinomas (at protein level).
Type: Primary
Antigen: Mesothelin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human