Anti-GRIN2B Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9373
BSBTPB9373EA 552 EUR
BSBTPB9373
Anti-GRIN2B Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Glutamate receptor ionotropic, NMDA 2B(GRIN2B) detection. Tested with WB in Human;Mouse;Rat.

The N-methyl-D-aspartate receptor 2B (NMDAR2B), also names as GRIN2B. The sequence of the predicted 1,484-amino acid human protein is 98% and 96% identical to the sequences of the rat and mouse Nmdar2b proteins, respectively. Nmdar2B gene is located on mouse chromosome 6 between Rho and Ly49 centromerically and Glb telomerically. Mapping of the human NMDAR2B receptor subunit gene (GRIN2B) to chromosome 12p12 overexpression of NMDA receptor 2B (NR2B) in the forebrains of transgenic mice leads to enhanced activation of NMDA receptors, facilitating synaptic potentiation in response to stimulation at 10-100 Hz.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for NMDAR2B is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: GRIN2C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR