Rabbit IgG polyclonal antibody for Peroxiredoxin-5, mitochondrial(PRDX5) detection. Tested with WB, IHC-P, ICC in Human;Mouse;Rat.
PRDX5 (peroxiredoxin 5) also known as AOEB166, ACR1,B166, MGC117264, MGC142283, MGC142285, PLP, PMP20, PRDX6, PRXV, SBBI10, is a member of the peroxiredoxin family and may play an antioxidant protective role in various tissues under nonpathologic conditions and during inflammatory processes. The PRDX5 gene is mapped to11q13.1. PRDX5 displays mitochondrial presequence features and has 3 cysteines implicated in antioxidant activity and a C-terminal SQL peroxisomal targeting sequence. Northern blot analysis revealed ubiquitous expression of a 1.0-kb PRDX5 transcript in tissues and cell lines. Functional analysis showed that PRDX5 has antioxidant activity equivalent to that of CAT. While PRDX5 was localized to fibroblasts in normal tendon, it was localized to fibroblasts and endothelial cells in degenerative tendon. PRDX5 mRNA and protein levels increased at 12 hours, and the increase in PRDX5 expression correlated with reduced peroxide levels. PRDX5 plays a protective role against oxidative stress in human cartilage.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
WB: The detection limit for Peroxiredoxin 5 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: PTEN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
, Human