Anti-SMAD1/2/3/4/5 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9395
BSBTPB9395EA 552 EUR
BSBTPB9395
Anti-SMAD1/2/3/4/5 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Mothers against decapentaplegic homolog 1/2/3/4/5(SMAD1/2/3/4/5) detection. Tested with WB in Human;Mouse;Rat.

SMADs are proteins that modulate the activity of transforming growth factor beta ligands. The SMADs, often in complex with other SMADs/CoSMAD, act as transcription factors that regulate the expression of certain genes. It was concluded that targeted ubiquitination of SMADs may serve to control both embryonic development and a wide variety of cellular responses to TGF-beta signals. R-Smads or receptor regulated Smads are a class of proteins that include SMAD1, SMAD2, SMAD3, SMAD5, and SMAD8. In response to signals by the TGF-? superfamily of ligands these proteins associate with receptor kinases and are phosphorylated at an SSXS motif at their extreme C-terminus. These proteins then typically bind to the common mediator Smad or co-SMAD SMAD4.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for SMAD1/2/3/4/5 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: SMAD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR