Anti-TNF Receptor II Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9514
BSBTPB9514EA 552 EUR
BSBTPB9514
Anti-TNF Receptor II Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Tumor necrosis factor receptor superfamily member 1B (TNFRSF1B) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: TNF Receptor II
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR