Anti-TRAF2 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9516
BSBTPB9516EA 552 EUR
BSBTPB9516
Anti-TRAF2 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for TNF receptor-associated factor 2(TRAF2) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: TRAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR