Anti-Cdk7 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9536
BSBTPB9536EA 552 EUR
BSBTPB9536
Anti-Cdk7 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Cyclin-dependent kinase 7(CDK7) detection. Tested with WB in Human.

Type: Primary
Antigen: Cdk7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

TECHNISCHE DATEN

Antigen-Symbol CDK7
Environmentally Preferable
Antigen-Name Cyclin-dependent kinase 7
Antigen-Synonyme CAK1|MO15|HCAK|CDKN7|STK1|p39MO15
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Gen-ID 1022
Isotyp IgG
Western blot Yes
Kreuzadsorption No

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR