Anti-Cdk9 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9537
BSBTPB9537EA 552 EUR
BSBTPB9537
Anti-Cdk9 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Cyclin-dependent kinase 9(CDK9) detection. Tested with WB, IHC-P in Human;Rat.

Type: Primary
Antigen: Cdk9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR