Anti-EBAG9 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9553
BSBTPB9553EA 552 EUR
BSBTPB9553
Anti-EBAG9 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Receptor-binding cancer antigen expressed on SiSo cells(EBAG9) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: EBAG9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

TECHNISCHE DATEN

Antigen-Symbol EBAG9
Environmentally Preferable
Antigen-Name Estrogen receptor binding site associated, antigen, 9
Antigen-Synonyme PDAF|EB9
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 9166
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
Kreuzadsorption No

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR