Anti-SP4 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9558
BSBTPB9558EA 552 EUR
BSBTPB9558
Anti-SP4 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Transcription factor Sp4(SP4) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: SP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR