Anti-SPARC Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9559
BSBTPB9559EA 552 EUR
BSBTPB9559
Anti-SPARC Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for SPARC(SPARC) detection. Tested with WB in Human.

Type: Primary
Antigen: SPARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR