Anti-G-CSF Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9563
BSBTPB9563EA 552 EUR
BSBTPB9563
Anti-G-CSF Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Granulocyte colony-stimulating factor(CSF3) detection. Tested with WB, ELISA in Human.

Type: Primary
Antigen: G-CSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR