Anti-FGF2 Antibody

Lieferant: Boster Bio
PB9587
BSBTPB9587EA 552 EUR
BSBTPB9587
Anti-FGF2 Antibody
Antikörper
Rabbit IgG polyclonal antibody for Fibroblast growth factor 2(FGF2) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: FGF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR