Anti-KIN Antibody

Lieferant: Boster Bio
PB9607
BSBTPB9607EA 552 EUR
BSBTPB9607
Anti-KIN Antibody
Antikörper
Rabbit IgG polyclonal antibody for DNA/RNA-binding protein KIN17(KIN) detection. Tested with WB, IHC-P in Human;Mouse.

Type: Primary
Antigen: KIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR