Anti-Lamin B1 Antibody

Lieferant: Boster Bio
PB9611
BSBTPB9611EA 552 EUR
BSBTPB9611
Anti-Lamin B1 Antibody
Antikörper
Rabbit IgG polyclonal antibody for Lamin-B1(LMNB1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: Lamin B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR