Anti-KCNA3 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PB9650
BSBTPB9650EA 552 EUR
BSBTPB9650
Anti-KCNA3 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Potassium voltage-gated channel subfamily A member 3(KCNA3) detection. Tested with WB in Human;Mouse;Rat.
Potassium voltage-gated channel, shaker-related subfamily, member 3, also known as KCNA3 or Kv1.3, is a protein that in humans is encoded by the KCNA3 gene. This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. This member contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. It belongs to the delayed rectifier class, members of which allow nerve cells to efficiently repolarize following an action potential. It plays an essential role in T-cell proliferation and activation. This gene appears to be intronless and it is clustered together with KCNA2 and KCNA10 genes on chromosome 1. And Kv1.3 has been reported to be expressed in the inner mitochondrial membrane in lymphocytes. The apoptotic protein Bax has been suggested to insert into theouter mitochondrial membrane and occlude the pore of Kv1.3 via a lysine residue. Thus, Kv1.3 modulation may be one of many mechanisms that contribute to apoptosis.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.