Anti-ADRBK2 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9682
BSBTPB9682EA 552 EUR
BSBTPB9682
Anti-ADRBK2 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Beta-adrenergic receptor kinase 2(ADRBK2) detection. Tested with WB in Human.

Type: Primary
Antigen: ADRBK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR