Anti-ARID1A Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9685
BSBTPB9685EA 552 EUR
BSBTPB9685
Anti-ARID1A Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for AT-rich interactive domain-containing protein 1A(ARID1A) detection. Tested with WB in Human.

Type: Primary
Antigen: ARID1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR