Anti-CXCL10 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9695
BSBTPB9695EA 552 EUR
BSBTPB9695
Anti-CXCL10 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for C-X-C motif chemokine 10(CXCL10) detection. Tested with WB, ELISA in Human.

Type: Primary
Antigen: CXCL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR