Anti-LIMK2 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9717
BSBTPB9717EA 552 EUR
BSBTPB9717
Anti-LIMK2 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for LIM domain kinase 2(LIMK2) detection. Tested with WB in Human;Mouse.

Type: Primary
Antigen: LIMK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR