Anti-MEK3 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9763
BSBTPB9763EA 552 EUR
BSBTPB9763
Anti-MEK3 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Dual specificity mitogen-activated protein kinase kinase 3(MAP kinase kinase 3/MAPKK 3)(MAP2K3) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: MEK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR