Anti-NR3C2 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9765
BSBTPB9765EA 552 EUR
BSBTPB9765
Anti-NR3C2 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Mineralocorticoid receptor(NR3C2) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: NR3C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR