Anti-PDGFRA Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9771
BSBTPB9771EA 552 EUR
BSBTPB9771
Anti-PDGFRA Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Platelet-derived growth factor receptor alpha(PDGFRA) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: PDGFRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR