Anti-CD45 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9785
BSBTPB9785EA 552 EUR
BSBTPB9785
Anti-CD45 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Receptor-type tyrosine-protein phosphatase C(PTPRC) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: CD45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR