Anti-ROC1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9798
BSBTPB9798EA 552 EUR
BSBTPB9798
Anti-ROC1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for E3 ubiquitin-protein ligase RBX1(RBX1) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: ROC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR