Anti-RAP1A Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9816
BSBTPB9816EA 552 EUR
BSBTPB9816
Anti-RAP1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAP1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR