Anti-SULT2A1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9821
BSBTPB9821EA 552 EUR
BSBTPB9821
Anti-SULT2A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SULT2A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

TECHNISCHE DATEN

Antigen-Symbol SULT2A1
Antigen-Name Sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1
Environmentally Preferable
Antigen-Synonyme hSTa|ST2|ST2A3|DHEAS|STD|ST2A1|HST|DHEA-ST
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 6822
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
Kreuzadsorption No

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR