Anti-UBE2Q2 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9839
BSBTPB9839EA 552 EUR
BSBTPB9839
Anti-UBE2Q2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UBE2Q2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR