Anti-UPF3B Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9843
BSBTPB9843EA 552 EUR
BSBTPB9843
Anti-UPF3B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UPF3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR