Anti-CTSK Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9856
BSBTPB9856EA 552 EUR
BSBTPB9856
Anti-CTSK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CTSK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR