Anti-CAP1 Antibody

Lieferant: Boster Bio
PB9923
BSBTPB9923EA 552 EUR
BSBTPB9923
Anti-CAP1 Antibody
Antikörper
Rabbit IgG polyclonal antibody for Adenylyl cyclase-associated protein 1(CAP1) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: CAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR