Anti-ABCB10 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PB9971
BSBTPB9971EA 552 EUR
BSBTPB9971
Anti-ABCB10 Rabbit Polyclonal Antibody
Antikörper
ABCB10, also known as M-ABC2, is expressed as a 60-kD nonglycosylated mitochondrial membrane protein. This ABCB10 gene is mapped to 1q42.13. The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. And ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The function of this mitochondrial protein is unknown.
1. Allikmets, R., Gerrard, B., Glavac, D., Ravnik-Glavac, M., Jenkins, N. A., Gilbert, D. J., Copeland, N. G., Modi, W., Dean, M.Characterization and mapping of three new mammalian ATP-binding transporter genes from an EST database. Mammalian Genome 6: 114-117, 1995.
2. Chen, W., Paradkar, P. N., Li, L., Pierce, E. L., Langer, N. B., Takahashi-Makise, N., Hyde, B. B., Shirihai, O. S., Ward, D. M., Kaplan, J., Paw, B. H. Abcb10 physically interacts with mitoferrin-1 (Slc25a37) to enhance its stability and function in the erythroid mitochondria. Proc. Nat. Acad. Sci. 106: 16263-16268, 2009.
3. Zhang, F., Hogue, D. L., Liu, L., Fisher, C. L., Hui, D., Childs, S., Ling, V. M-ABC2, a new human mitochondrial ATP-binding cassette membrane protein. FEBS Lett. 478: 89-94, 2000.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.