Anti-BCR Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PB9988
BSBTPB9988EA 552 EUR
BSBTPB9988
Anti-BCR Rabbit Polyclonal Antibody
Antikörper
The breakpoint cluster region protein (BCR) is a protein that in humans is encoded by the BCR gene. A reciprocal translocation between chromosomes 22 and 9 produces the Philadelphia chromosome, which is often found in patients with chronic myelogenous leukemia. The chromosome 22 breakpoint for this translocation is located within the BCR gene. The translocation produces a fusion protein which is encoded by sequence from both BCR and ABL, the gene at the chromosome 9 breakpoint. Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for p21rac. Two transcript variants encoding different isoforms have been found for this gene.
1. "Entrez Gene: Breakpoint cluster region".
2. Puil L, Liu J, Gish G, Mbamalu G, Bowtell D, Pelicci PG, Arlinghaus R, Pawson T (February 1994). "Bcr-Abl oncoproteins bind directly to activators of the Ras signalling pathway". EMBO J. 13 (4): 764–73.
3. Pendergast AM, Muller AJ, Havlik MH, Maru Y, Witte ON (July 1991). "BCR sequences essential for transformation by the BCR-ABL oncogene bind to the ABL SH2 regulatory domain in a non-phosphotyrosine-dependent manner".Cell 66 (1): 161–71.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.