Anti-Occludin Polyclonal Antibody

Lieferant: BosterBio
RP1057
BSBTRP1057EA 552 EUR
BSBTRP1057
Anti-Occludin Polyclonal Antibody
Antikörper
Polyclonal antibody for Occludin/OCLN detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. Occludin/OCLN information: Molecular Weight: 59144 MW; Subcellular Localization: Membrane; Multi-pass membrane protein. Cell junction, tight junction; Tissue Specificity: Localized at tight junctions of both epithelial and endothelial cells. Highly expressed in kidney. Not detected in testis.

Type: Primary
Antigen: Occludin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR