Anti-PKC Delta Polyclonal Antibody

Lieferant: Boster Bio
RP1058
BSBTRP1058EA 552 EUR
BSBTRP1058
Anti-PKC Delta Polyclonal Antibody
Antikörper
Polyclonal antibody for PKC delta/PRKCD detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. PKC delta/PRKCD information: Molecular Weight: 77505 MW; Subcellular Localization: Cytoplasm. Cytoplasm, perinuclear region. Nucleus. Endoplasmic reticulum. Mitochondrion. Cell membrane; Peripheral membrane protein.

Type: Primary
Antigen: PKC Delta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR