Anti-HGF Polyclonal Antibody

Lieferant: Boster Bio
RP1062
BSBTRP1062EA 552 EUR
BSBTRP1062
Anti-HGF Polyclonal Antibody
Antikörper
Polyclonal antibody for HGF detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Mouse. HGF information: Molecular Weight: 82945 MW; .

Type: Primary
Antigen: HGF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR