Anti-NRIP1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
RP1095
BSBTRP1095EA 552 EUR
BSBTRP1095
Anti-NRIP1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Nuclear receptor-interacting protein 1(NRIP1) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: NRIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR