Anti-CACYBP Antibody

Lieferant: Boster Bio
RP1104
BSBTRP1104EA 552 EUR
BSBTRP1104
Anti-CACYBP Antibody
Antikörper
Rabbit IgG polyclonal antibody for Calcyclin-binding protein(CACYBP) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: CACYBP
Clonality: Polyclonal
Clone: Clone
Conjugation: Conjugation
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR