Anti-BDNF Chicken Polyclonal Antibody

Lieferant: Biosensis
C-1517-500
BSENC-1517-500EA 321 EUR
BSENC-1517-500
Anti-BDNF Chicken Polyclonal Antibody
Antikörper
BDNF belongs to the neurotrophin family and regulates the survival and differentiation of neurons during development. The alterations in BDNF expression induced by various kinds of brain insult including stress, ischemia, seizure activity and hypoglycemia, may contribute to some pathologies such as depression, epilepsy, Alzheimer's, and Parkinson's disease. Microglia release BDNF that may contribute to neuroinflammation and neuropathic pain. FUNCTION: Promotes the survival of neuronal populations that are all located either in the central nervous system or directly connected to it. Major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. The versatility of BDNF is emphasized by its contribution to a range of adaptive neuronal responses including long-term potentiation (LTP), long-term depression (LTD), certain forms of short-term synaptic plasticity, as well as homeostatic regulation of intrinsic neuronal excitability. SUBUNIT: Monomers and homodimers. Binds to NTRK2/TRKB. SUBCELLULAR LOCATION: Secreted protein. POst translation modification: Converted into mature BDNF by plasmin (PLG). SIMILARITY: Belongs to the NGF-beta family.

The antibody specifically reacts with BDNF protein. The antibody has been successfully tested in ELISA. The antibody has not been tested in western blot or IHC. BDNF is highly conserved in human, mouse, and rat, as well as other mammalian species so cross reactivity to BDNF from other mammals is expected.
Application Information:
ELISA. Suggested dilution at 1:500 to 1:2,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: BDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human BDNF peptides consisting of amino acids 73-87 and 194-208.
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR