Anti-Rhodopsin Chicken Polyclonal Antibody

Lieferant: Biosensis
C-1521-500
BSENC-1521-500EA 321 EUR
BSENC-1521-500
Anti-Rhodopsin Chicken Polyclonal Antibody
Antikörper
Rod shaped photoreceptor cells that are required for image-forming vision at low light intensity and for photoreceptor cell viability after birth (ref: SWISSPROT).

The antibody reacts in wester blots in mouse brain extracts with a prominent band ~36kDa cooresponding to Rhodopsin. The antibody has not been tested in IHC.
Application Information:
WB. Suggested dilution at 1:500 to 1:2,000. To minimise background staining, a higher concentration of detergent (such as Tween-20) is suggested in the dilution and washing steps. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Rhodopsin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Peptides cooresponding to amino acids 14-26 of Mouse Opsin 1 and amino acids 18-30 of Tiger finch opsin1.
Host: Chicken
Isotype: IgY
Reactivity: Mouse, Rat, Tiger Finch, Zebra Finch
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR