Anti-CNTF Chicken Polyclonal Antibody

Lieferant: Biosensis
C-1526-500
BSENC-1526-500EA 321 EUR
BSENC-1526-500
Anti-CNTF Chicken Polyclonal Antibody
Antikörper
CNTF is a survival promoting factor for different types of neurons in vitro and in vivo. The essential structural features for the biological function of human CNTF were investigated by Thier, M. et al. They showed that deletion of 14 N-terminal and 18 C-terminal amino acids significantly increased bioactivity compared to wild-type CNTF. FUNCTION: CNTF is a survival factor for various neuronal cell types. Seems to prevent the degeneration of motor axons after axotomy. SUBUNIT: Homodimer. SUBCELLULAR LOCATION: Cytoplasm. TISSUE SPECIFICITY: Nervous system. PHARMACEUTICAL: CNTF is being tested under the name Axokine by Regeneron Pharmaceuticals for treatment of human motor neuron diseases, such as amyotrophic lateral sclerosis (ALS). As it induces substantial weight loss, preferentially of fat as opposed to lean body mass, it is being used for obesity treatment. SIMILARITY: Belongs to the CNTF family.

This antibody detects CNTF from human, rat, and mouse extracts in western blot and ELISA. Because of homology it is expected to react with other mammalian species as well. The antibody has not been tested in IHC
Application Information:
WB and ELISA at a suggested dilution of 1:500 to 1:2,000. A titration between 1:50 to 1:500 is suggested for IHC. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: CNTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Maps to amino acids 11-25 and 186-200 of Human CNTF protein
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR