Anti-TrkA Mouse Monoclonal Antibody [clone: BS292]

Lieferant: Biosensis
M-1719-100
BSENM-1719-100EA 312 EUR
BSENM-1719-100
Anti-TrkA Mouse Monoclonal Antibody [clone: BS292]
Antikörper
TrkA is a member of the neurotrophic tyrosine kinase receptor family. It is a membrane-bound receptor that upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. TrkA is required for high-affinity binding to nerve growth factor (NGF), neurotrophin-3 and neurotrophin-4/5 but not brain-derived neurotrophic factor (BDNF). TrkA leads to cell differentiations and may play a role in specifying sensory neuron subtypes. It has a crucial role in the development and function of the nociceptive reception system as well as establishment of thermal regulation via sweating. SUBUNIT: Exists in a dynamic equilibrium between monomeric (low affinity) and dimeric (high affinity) structures. SUBCELLULAR LOCATION: Cell membrane; single-pass type I membrane protein. Endocytosed to the endosomes upon treatment of cells with NGF. ALTERNATIVE PRODUCTS: 2 named isoforms produced by alternative splicing. Both isoforms have similar biological properties. TISSUE SPECIFICITY: Isoform TrkA-II is primarily expressed in neuronal cells. Isoform TrkA-I is found in non-neuronal tissues. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutalating behaviour, mental retardation and cancer (Reference: www.uniprot.com).

WB: Western blotting: 1-3 µg/mL, antibody is suitable for reduced (with DTT or ?-mercaptoethanol) and non-reduced samples. Denatured but not reduced samples provides cleaner blot signals as demonstrated in our photographs.

Immunocytochemistry: 1-2 µg/mL. Antibody works on 4% formaldehyde-fixed cells. Note that cells require a permeabilization step, because the antibody detects a cytoplasmic epitope of TrkA.

Other applications not yet tested. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TrkA
Clonality: Monoclonal
Clone: BS292
Conjugation: Unconjugated
Epitope: A synthetic peptide from the intracellular cytoplasmic domain of human TrkA (C-HGPDAKLLAGGE, aa: 604-615) has been used as the immunogen.
Sequence
Host: Mouse
Isotype: IgG3, k-light chain
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR